missing translation for 'onlineSavingsMsg'
Learn More

3Alpha,7Alpha,12Alpha-Trihydroxycoprostanic Acid-d3, TRC

Product Code. 30456948
Click to view available options
Quantity:
0.5 mg
Unit Size:
0.5mg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30456948

Brand: Toronto Research Chemicals TRCT7951570.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3alpha,7alpha,12alpha-Trihydroxycoprostanic Acid-d3
CAS 338976-79-7
Molecular Formula C27H43D3O5
Quantity 0.5 mg
Synonym (3α,5β,7α,12α)-3,7,12-Trihydroxy-cholestan-26-oic-27,27,27-d3 Acid, (3α,5β,7α,12α)-3,7,12-Trihydroxycholestan-26-oic Acid-d3, 3α,7α,12α-Trihydroxy-5β-cholestan-26-oic Acid-d3, 3α,7α,12α-Trihydroxycoprostanic Acid-d3, 5β-Cholestane-3α,7α,12α-triol-26-oic Acid-d3, Coprocholic Acid-d3, Trihydroxycoprostanic Acid-d3
InChI Formula InChI=1S/C27H46O5/c1-15(6-5-7-16(2)25(31)32)19-8-9-20-24-21(14-23(30)27(19,20)4)26(3)11-10-18(28)12-17(26)13-22(24)29/h15-24,28-30H,5-14H2,1-4H3,(H,31,32)/t15-,16?,17+,18-,19-,20+,21+,22-,23+,24+,26+,27-/m1/s1/i2D3
SMILES O[C@@H]1CC[C@@]2(C)[C@@](C[C@@H](O)[C@]3([H])[C@]2([H])C[C@H](O)[C@@]4(C)[C@]3(CC[C@@]4([C@H](C)CCCC(C([2H])([2H])[2H])C(O)=O)[H])[H])([H])C1
Molecular Weight (g/mol) 453.67
Formula Weight 453.3534
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.