missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-1,4-Dibromo-2-butene-d6, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | (2 E)-1,4-Dibromo-2-butene-D6 |
| CAS | 118886-18-3 |
| Molecular Formula | C4 D6 Br2 |
| Quantity | 0.5 g |
| Synonym | 2-Butene-1,1,2,3,4,4-d6, 1,4-dibromo-, (2 E)- (9 CI), 2-Butene-1,1,2,3,4,4-d6, 1,4-dibromo-, (E)- (ZCI), trans-1,4-Dibromo-2-butene-d6, (2 E)-1,4-Dibromo-2-butene-D6, (2 E)-1,4-Dibromo-2-butene-1,1,2,3,4,4-d6 |
| InChI Formula | InChI=1 S/C4H6Br2/c5-3-1-2-4-6/h1-2 H,3-4H2/b2-1+/i1D,2 D,3D2,4D2 |
| SMILES | [2 H]\C(=C(\[2 H])/C([2 H])([2 H])Br)\C([2 H])([2 H])Br |
| IUPAC Name | (E)-1,4-dibromo-1,1,2,3,4,4-hexadeuteriobut-2-ene |
| Molecular Weight (g/mol) | 219.935 |
| Formula Weight | 217.921 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction