Learn More
Tetramethylsilane-d12, CDN
Product Code. 30321230
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30321230
Brand: CDN CDND04850.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Tetramethylsilane-D12
CAS 18145-38-5
Molecular Formula C4D12Si
Quantity 0.5 g
Synonym Tetra(methyl-d3)silane, Tetrakis(trideuteromethyl)silane, Tetramethylsilane-d12
InChI Formula InChI=1 S/C4H12Si/c1-5(2,3)4/h1-4H3/i1D3,2D3,3D3,4D3
SMILES [2 H]C([2 H])([2 H])[Si](C([2 H])([2 H])[2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H]
IUPAC Name tetrakis(methyl-d3)silane
Molecular Weight (g/mol) 100.3
Formula Weight 100.15 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity