missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Tetrahydro-2-furoic-d7 Acid, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 2-Tetrahydrofurancarboxylic acid d7 |
| CAS | 1219798-42-1 |
| Molecular Formula | C5 2H7 H O3 |
| Quantity | 0.5 g |
| Synonym | 2-Tetrahydrofurancarboxylic acid D7, (±)-Tetrahydro-2-furoic-d7 acid, Tetrahydro-2-furancarboxylic-2,3,3,4,4,5,5-d7 acid, 2-Tetrahydrofurancarboxylic-d7 acid, Tetrahydro-2,3,3,4,4,5,5-heptadeuterio-2-furancarboxylic acid |
| InChI Formula | InChI=1 S/C5H8O3/c6-5(7)4-2-1-3-8-4/h4H,1-3H2,(H,6,7)/i1D2,2D2,3D2,4 D |
| SMILES | [2 H]C1([2 H])OC([2 H])(C(=O)O)C([2 H])([2 H])C1([2 H])[2 H] |
| IUPAC Name | 2,3,3,4,4,5,5-heptadeuteriooxolane-2-carboxylic acid |
| Molecular Weight (g/mol) | 123.16 |
| Formula Weight | 123.0913 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction