missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Octadecanoate-d35, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Sodium Octadecanoate-D35 |
| CAS | 1219794-97-4 |
| Molecular Formula | C18 D35 O2 . Na |
| Quantity | 0.5 g |
| Synonym | Octadecanoic-d35 acid sodium salt, Stearic-d35 acid sodium salt, Sodium stearate-d35, Sodium octadecanoate-D35 |
| InChI Formula | InChI=1 S/C18H36O2.Na/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20;/h2-17H2,1H3,(H,19,20);/q;+1/p-1/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2,16D2,17D2; |
| SMILES | [Na+].[2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C(=O)[O-] |
| IUPAC Name | sodium;2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16,17,17,18,18,18-pentatriacontadeuteriooctadecanoate |
| Molecular Weight (g/mol) | 341.675 |
| Formula Weight | 341.473 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction