missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium 2-Keto-3-methyl-d3-butyrate-3,4,4,4-d4, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.50g
1g
Specifications
Specifications
| Chemical Name or Material | Sodium 3-Methyl-2-oxobutanoate-D7 |
| CAS | 2483831-46-3 |
| Molecular Formula | C5 D7 O3 . Na |
| Quantity | 0.5 g |
| Synonym | Butanoic-3,4,4,4-d4 acid, 3-(methyl-d3)-2-oxo-, sodium salt (1:1) (ACI), Sodium α-ketoisovalerate-d7, Sodium 3-methyl-2-oxobutanoate-D7, Sodium dimethylpyruvate-d7, Sodium α-ketovaline-d7, 3-(Methyl-d3)-2-oxobutyric-3,4,4,4-d4 acid sodium salt, 3-(Methyl-d3)-2-oxobutanoic-3,4,4,4-d4 acid sodium salt |
| InChI Formula | InChI=1 S/C5H8O3.Na/c1-3(2)4(6)5(7)8;/h3H,1-2H3,(H,7,8);/q;+1/p-1/i1D3,2D3,3 D; |
| SMILES | [Na+].[2 H]C([2 H])([2 H])C([2 H])(C(=O)C(=O)[O-])C([2 H])([2 H])[2 H] |
| IUPAC Name | sodium;3,4,4,4-tetradeuterio-2-oxo-3-(trideuteriomethyl)butanoate |
| Molecular Weight (g/mol) | 145.14 |
| Formula Weight | 145.073 g/mol |
| Show More |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion