Learn More
Pyridine-2,6-d2, CDN
Product Code. 30324188
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30324188
Brand: CDN CDND03670.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Pyridine-2,6-D2
CAS 17265-96-2
Molecular Formula C5H3D2N
Quantity 0.5 g
Synonym Pyridine-2,6-d2 (6 CI, 7 CI, 8 CI, 9 CI, ACI), 2,6-Dideuteropyridine, Pyridine-2,6-D2
InChI Formula InChI=1 S/C5H5N/c1-2-4-6-5-3-1/h1-5 H/i4D,5 D
SMILES [2 H]C1=CC=CC([2 H])=N1
IUPAC Name pyridine-2,6-d2
Molecular Weight (g/mol) 81.11
Formula Weight 81.05 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity