missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenyl-d5-acetic Acid, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Phenylacetic Acid D5 (phenyl-D5) |
| CAS | 104182-98-1 |
| Molecular Formula | C8 D5 H3 O2 |
| Quantity | 0.5 g |
| Synonym | Benzene-2,3,4,5,6-d5-acetic acid (ACI), Benzene-d5-acetic acid (9 CI), Phenyl-d5-acetic acid, Phenylacetic acid-d5, Phenylacetic acid-d5 |
| InChI Formula | InChI=1 S/C8H8O2/c9-8(10)6-7-4-2-1-3-5-7/h1-5 H,6H2,(H,9,10)/i1D,2 D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(CC(=O)O)c([2 H])c1[2 H] |
| IUPAC Name | 2-(2,3,4,5,6-pentadeuteriophenyl)acetic acid |
| Molecular Weight (g/mol) | 141.1787 |
| Formula Weight | 141.0838 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction