Learn More
p-Toluidine-d7 (NH2), CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.50g
1g
Specifications
Specifications
| Chemical Name or Material | 4-Toluidine-d7 |
| CAS | 68693-08-3 |
| Molecular Formula | C7 D7 H2 N |
| Quantity | 0.5 g |
| Synonym | Benzen-2,3,5,6-d4-amine, 4-(methyl-d3)- (9 CI, ACI), 4-(Methyl-d3)benzen-2,3,5,6-d4-amine (ACI), 4-Toluidine-d7, p-Toluidine-d7, 4-Methylaniline-D7 |
| InChI Formula | InChI=1 S/C7H9N/c1-6-2-4-7(8)5-3-6/h2-5 H,8H2,1H3/i1D3,2 D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c(c([2 H])c([2 H])c1N)C([2 H])([2 H])[2 H] |
| IUPAC Name | 2,3,5,6-tetradeuterio-4-(trideuteriomethyl)aniline |
| Molecular Weight (g/mol) | 114.196 |
| Formula Weight | 114.117 g/mol |
| Show More |