Learn More
Octadecanoic-2,2-d2 Acid, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.50g
1g
Specifications
Specifications
| Chemical Name or Material | Octadecanoic-2,2-D2 Acid |
| CAS | 19905-58-9 |
| Molecular Formula | C18 D2 H34 O2 |
| Quantity | 0.5 g |
| Synonym | Octadecanoic-2,2-d2 acid (9 CI, ACI), Stearic-2,2-d2 acid (7 CI, 8 CI), 2,2-Dideuterostearic acid, Octadecanoic-2,2-D2 acid |
| InChI Formula | InChI=1 S/C18H36O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h2-17H2,1H3,(H,19,20)/i17D2 |
| SMILES | [2 H]C([2 H])(CCCCCCCCCCCCCCCC)C(=O)O |
| IUPAC Name | 2,2-dideuteriooctadecanoic acid |
| Molecular Weight (g/mol) | 286.49 |
| Formula Weight | 286.284 g/mol |
| Show More |