missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Propyl-2,2,3,3,3-d5-triphenylphosphonium Bromide, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | n-Propyltriphenylphosphonium-d5 Bromide |
| CAS | 1703757-22-5 |
| Molecular Formula | C21 D5 H17 P . Br |
| Quantity | 0.5 g |
| Synonym | Triphenylpropylphosphonium-d5 Bromide, Bromo(propyl)triphenylphosphorane-d5, NSC 50539-d5, Propyltriphenylphosphonium-d5 Bromide; |
| InChI Formula | InChI=1 S/C21H22P.BrH/c1-2-18-22(19-12-6-3-7-13-19,20-14-8-4-9-15-20)21-16-10-5-11-17-21;/h3-17 H,2,18H2,1H3;1 H/q+1;/p-1/i1D3,2D2; |
| SMILES | [Br-].[2 H]C([2 H])([2 H])C([2 H])([2 H])C[P+](c1ccccc1)(c2ccccc2)c3ccccc3 |
| IUPAC Name | 2,2,3,3,3-pentadeuteriopropyl(triphenyl)phosphanium;bromide |
| Molecular Weight (g/mol) | 390.3079 |
| Formula Weight | 389.0956 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction