missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Octylbenzene-2,3,4,5,6-d5, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Octylbenzene-D5 (phenyl-D5) |
| CAS | 1219799-23-1 |
| Molecular Formula | C14 D5 H17 |
| Quantity | 0.5 g |
| Synonym | Benzene-1,2,3,4,5-d5, 6-octyl- (ACI), 6-Octylbenzene-1,2,3,4,5-d5 (ACI), Octylbenzene-d5, Octylbenzene-D5 (phenyl-D5), 1-Octylbenzene-2,3,4,5,6-d5, 1-(Phenyl-d5)octane, n-Octylbenzene-2,3,4,5,6-d5 |
| InChI Formula | InChI=1 S/C14H22/c1-2-3-4-5-6-8-11-14-12-9-7-10-13-14/h7,9-10,12-13 H,2-6,8,11H2,1H3/i7D,9 D,10 D,12 D,13 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(CCCCCCCC)c([2 H])c1[2 H] |
| IUPAC Name | 1,2,3,4,5-pentadeuterio-6-octylbenzene |
| Molecular Weight (g/mol) | 195.355 |
| Formula Weight | 195.204 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction