Learn More
N,N-Dimethylethanolamine-d10 (OH), CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.50g
Specifications
Specifications
| Chemical Name or Material | Deanol-D10 |
| CAS | 2567957-94-0 |
| Molecular Formula | C4 D10 H N O |
| Quantity | 0.5 g |
| Synonym | N,N-Dimethylethanolamine-d10 (OH), 2-[Di(methyl-d3)amino]ethanol-1,1,2,2-d4, N,N-Di(methyl-d3)ethanolamine-1,1,2,2-d4, Deanol-d10, Deanol-D10, 2-Dimethylaminoethanol-d10 |
| InChI Formula | InChI=1 S/C4H11NO/c1-5(2)3-4-6/h6H,3-4H2,1-2H3/i1D3,2D3,3D2,4D2 |
| SMILES | [2 H]C([2 H])([2 H])N(C([2 H])([2 H])[2 H])C([2 H])([2 H])C([2 H])([2 H])O |
| IUPAC Name | 2-[bis(trideuteriomethyl)amino]-1,1,2,2-tetradeuterioethanol |
| Molecular Weight (g/mol) | 99.198 |
| Formula Weight | 99.147 g/mol |
| Show More |