Learn More
n-Hexyl-d13 Chloroformate, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Hexyl Chloroformate D13 |
| CAS | 2708278-38-8 |
| Molecular Formula | C7 D13 Cl O2 |
| Quantity | 0.5 g |
| Synonym | n-Hexyl-d13 chloroformate, Carbonochloridic acid hexyl-d13 ester, Hexyl-d13 chlorocarbonate, Hexyl chloroformate-d13, Hexyl chloroformate D13 |
| InChI Formula | InChI=1 S/C7H13ClO2/c1-2-3-4-5-6-10-7(8)9/h2-6H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])OC(=O)Cl |
| IUPAC Name | 1,1,2,2,3,3,4,4,5,5,6,6,6-tridecadeuteriohexyl carbonochloridate |
| Molecular Weight (g/mol) | 177.71 |
| Formula Weight | 177.142 g/mol |
| Show More |