missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Butylamine-ND2 DCl, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 1-Butylamine-N,N-D2 Deuterochloride |
| CAS | 156235-87-9 |
| Molecular Formula | C4 D2 H9 N . Cl D |
| Quantity | 0.5 g |
| Synonym | Hydrochloric acid-d, compd. with 1-butanamine-d2 (1:1) (9 CI), 1-Butanamine-d2, hydrochloride-d (9 CI), 1-Butanamine-d2 hydrochloride-d, 1-Butanamine-N,N-d2 hydrochloride-d, 1-Butylamine-N,N-D2 deuterochloride, n-Butylamine-d2 deuterochloride, 1-Butanamine-ND2 deuterochloride |
| InChI Formula | InChI=1 S/C4H11N.ClH/c1-2-3-4-5;/h2-5H2,1H3;1 H/i/hD3 |
| SMILES | [2 H]Cl.[2 H]N([2 H])CCCC |
| Molecular Weight (g/mol) | 112.616 |
| Formula Weight | 112.085 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction