missing translation for 'onlineSavingsMsg'
Learn More
Learn More
m-Cresol-d3 (methyl-d3), CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | m-Cresol-d3 (methyl-d3) |
| CAS | 25026-32-8 |
| Molecular Formula | C7 D3 H5 O |
| Quantity | 0.5 g |
| Synonym | m-Cresol-d3 (methyl-d3), m-Cres-α,α,α-d3-ol (8 CI), 3-(Methyl-d3)phenol, m-Cresol-d3, 1-Hydroxy-3-(methyl-d3)benzene, 3-Cresol-d3, m-Hydroxytoluene-d3, Metacresol-D3 (methyl-D3) |
| InChI Formula | InChI=1 S/C7H8O/c1-6-3-2-4-7(8)5-6/h2-5,8 H,1H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])c1cccc(O)c1 |
| IUPAC Name | 3-(trideuteriomethyl)phenol |
| Molecular Weight (g/mol) | 111.1563 |
| Formula Weight | 111.0763 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction