missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Lysine-2,6,6-d3 HCl, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.50g
Specifications
Specifications
| Chemical Name or Material | L-Lysine-2,6,6-d3 HCl |
| CAS | 2330878-43-6 |
| Molecular Formula | C6 D3 H11 N2 O2 . H Cl |
| Quantity | 0.5 g |
| Synonym | L-Lysine-2,6,6-d3 HCl, L-Lysine-2,6,6-d3 hydrochloride, L-Lysine-2,6,6-d3 monohydrochloride, L-Lysine-2,6,6-D3 hydrochloride, (2 S)-2,6-Diamino-2,6,6-trideuteriohexanoic acid monohydrochloride |
| InChI Formula | InChI=1 S/C6H14N2O2.ClH/c7-4-2-1-3-5(8)6(9)10;/h5H,1-4,7-8H2,(H,9,10);1 H/t5-;/m0./s1/i4D2,5 D; |
| SMILES | Cl.[2 H]C([2 H])(N)CCC[C@]([2 H])(N)C(=O)O |
| IUPAC Name | (2 S)-2,6-diamino-2,6,6-trideuteriohexanoic acid;hydrochloride |
| Molecular Weight (g/mol) | 185.667 |
| Formula Weight | 185.101 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction