Learn More
Iodoethane-1-d1, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 1-Iodoethane-1-D |
| CAS | 3652-81-1 |
| Molecular Formula | C2 D H4 I |
| Quantity | 0.5 g |
| Synonym | Ethane-d, 1-iodo- (7 CI, 8 CI, 9 CI), [1-2H1]Ethyl iodide, 1-Iodoethane-d, 1-Iodoethane-1-d, Ethyl iodide-1-d, Ethyl-1-d1 iodide, 1-Iodoethane-1-D |
| InChI Formula | InChI=1 S/C2H5I/c1-2-3/h2H2,1H3/i2D |
| SMILES | [2 H]C(C)I |
| IUPAC Name | 1-deuterio-1-iodoethane |
| Molecular Weight (g/mol) | 156.9717 |
| Formula Weight | 156.9499 g/mol |
| Show More |