missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexanoic-6,6,6-d3 Acid, CDN
Click to view available options
Quantity:
0.1 g
0.5 g
Unit Size:
0.1g
0.50g
Specifications
Specifications
| Chemical Name or Material | Hexanoic-6,6,6-d3 Acid |
| CAS | 55320-69-9 |
| Molecular Formula | C6 D3 H9 O2 |
| Quantity | 0.5 g |
| Synonym | 1-Hexanoic-d3 Acid, 1-Pentane-d3-carboxylic Acid, Butyl-d3-acetic Acid, Caproic-d3 Acid, Capronic-d3 Acid, Hexanoic-6,6,6-d3 acid (9 CI), Hexanoic-6,6,6-d3 acid (9 CI), Hexanoic-6,6,6-d3 acid (9 CI), Hexanoic-6,6,6-d3 acid (9 CI), Hexanoic-6,6,6-d3 acid (9 CI), Hexanoic-6,6,6-d3 acid (9 CI), Hexanoic-6,6,6-d3 acid (9 CI), Hexanoic-6,6,6-d3 acid (9 CI), Hexanoic-6,6,6-d3 acid (9 CI), Hexanoic-6,6,6-d3 acid (9 CI), Hexanoic-6,6,6-d3 acid (9 CI), Hexanoic-6,6,6-d3 acid (9 CI), Hexanoic-6,6,6-d3 acid (9 CI) |
| InChI Formula | InChI=1 S/C6H12O2/c1-2-3-4-5-6(7)8/h2-5H2,1H3,(H,7,8)/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])CCCCC(=O)O |
| IUPAC Name | 6,6,6-trideuteriohexanoic acid |
| Molecular Weight (g/mol) | 119.18 |
| Formula Weight | 119.1026 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction