missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heptanoic-d13 Acid, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Heptanoic Acid-D13 |
| CAS | 130348-93-5 |
| Molecular Formula | C7 D13 H O2 |
| Quantity | 0.5 g |
| Synonym | Heptanoic-d13 acid (9 CI), Heptanoic-2,2,3,3,4,4,5,5,6,6,7,7,7-d13 acid, Heptanoic Acid-D13, Heptanoic acid-d13, Enanthic acid-d13 |
| InChI Formula | InChI=1 S/C7H14O2/c1-2-3-4-5-6-7(8)9/h2-6H2,1H3,(H,8,9)/i1D3,2D2,3D2,4D2,5D2,6D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C(=O)O |
| IUPAC Name | 2,2,3,3,4,4,5,5,6,6,7,7,7-tridecadeuterioheptanoic acid |
| Molecular Weight (g/mol) | 143.265 |
| Formula Weight | 143.181 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction