Learn More
Glycine-2,2-d2-N-t-BOC, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | N-[(1,1-Dimethylethoxy)carbonyl]glycine-2,2-d2 |
| CAS | 42492-65-9 |
| Molecular Formula | C7 D2 H11 N O4 |
| Quantity | 0.5 g |
| Synonym | Glycine-2,2-d2, N-[(1,1-dimethylethoxy)carbonyl]- (9 CI, ACI), N-[(1,1-Dimethylethoxy)carbonyl]glycine-2,2-d2 (ACI), 2-(tert-Butoxycarbonylamino)-2,2-dideuterio-acetic acid, N-(tert-Butoxycarbonyl)glycine-d2, N-(tert-Butoxycarbonyl)glycine-2,2-D2, N-Boc-glycine-2,2-d2, Glycine-2,2-d2-N-t-BOC |
| InChI Formula | InChI=1 S/C7H13NO4/c1-7(2,3)12-6(11)8-4-5(9)10/h4H2,1-3H3,(H,8,11)(H,9,10)/i4D2 |
| SMILES | [2 H]C([2 H])(NC(=O)OC(C)(C)C)C(=O)O |
| IUPAC Name | 2,2-dideuterio-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetic acid |
| Molecular Weight (g/mol) | 177.195 |
| Formula Weight | 177.097 g/mol |
| Show More |