missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl trans-Cinnamate-d5 (phenyl-d5), CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 3-Phenyl-2-propenoic Acid Ethyl Ester D5 (phenyl-D5) |
| CAS | 856765-68-9 |
| Molecular Formula | C11 2H5 H7 O2 |
| Quantity | 0.5 g |
| Synonym | 2-Propenoic acid, 3-(phenyl-d5)-, ethyl ester (9 CI), Ethyl trans-Cinnamate-d5, 3-(Phenyl-d5)-2-propenoic acid ethyl ester, Ethyl trans-cinnamate-d5 (phenyl-d5), 3-[2H5]Phenylacrylic acid ethyl ester, Ethyl cinnamate-d5, 3-Phenyl-2-propenoic acid ethyl est |
| InChI Formula | InChI=1 S/C11H12O2/c1-2-13-11(12)9-8-10-6-4-3-5-7-10/h3-9 H,2H2,1H3/i3D,4 D,5 D,6 D,7 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C=CC(=O)OCC)c([2 H])c1[2 H] |
| IUPAC Name | ethyl 3-(2,3,4,5,6-pentadeuteriophenyl)prop-2-enoate |
| Molecular Weight (g/mol) | 181.243 |
| Formula Weight | 181.115 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction