Learn More
Ethyl-2,2,2-d3-amine HCl, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Ethylamine Hydrochloride-d3 |
| CAS | 64585-12-2 |
| Molecular Formula | C2 D3 H4 N . Cl H |
| Quantity | 0.5 g |
| Synonym | Ethan-2,2,2-d3-amine, hydrochloride (9 CI), 2,2,2-Trideuterioethanamine hydrochloride, Ethan-2,2,2-d3-amine hydrochloride, Ethylamine-d3 hydrochloride, N-(Ethyl-2,2,2-d3)ammonium chloride, Ethylamine-2,2,2-D3 hydrochloride |
| InChI Formula | InChI=1 S/C2H7N.ClH/c1-2-3;/h2-3H2,1H3;1 H/i1D3; |
| SMILES | Cl.[2 H]C([2 H])([2 H])CN |
| IUPAC Name | 2,2,2-trideuterioethanamine;hydrochloride |
| Molecular Weight (g/mol) | 84.563 |
| Formula Weight | 84.053 g/mol |
| Show More |