missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Valine-2-d1, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | DL-Valine-2-D |
| CAS | 14246-21-0 |
| Molecular Formula | C5 D H10 N O2 |
| Quantity | 0.5 g |
| Synonym | Valine-2-d (9 CI, ACI), DL-Valine-2-d (ZCI), Valine-2-d, DL- (8 CI), DL-(α-2 H)Valine, 2-Amino-3-methylbutanoic-2-d acid, DL-Valine-2-D |
| InChI Formula | InChI=1 S/C5H11NO2/c1-3(2)4(6)5(7)8/h3-4 H,6H2,1-2H3,(H,7,8)/i4D |
| SMILES | [2 H]C(N)(C(C)C)C(=O)O |
| IUPAC Name | 2-amino-2-deuterio-3-methylbutanoic acid |
| Molecular Weight (g/mol) | 118.1525 |
| Formula Weight | 118.0853 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction