missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Phenylalanine-2-d1, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | DL-Phenylalanine-D (propanoic-2-D) |
| CAS | 14246-24-3 |
| Molecular Formula | C9 D H10 N O2 |
| Quantity | 0.5 g |
| Synonym | Phenylalanine-α-d (9 CI, ACI), DL-Phenylalanine-α-d (ZCI), Alanine-2-d, phenyl-, DL- (8 CI), DL-Phenylalanine-2-d1, DL-Phenylalanine-D (propanoic-2-D) |
| InChI Formula | InChI=1 S/C9H11NO2/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8 H,6,10H2,(H,11,12)/i8D |
| SMILES | [2 H]C(N)(Cc1ccccc1)C(=O)O |
| IUPAC Name | 2-amino-2-deuterio-3-phenylpropanoic acid |
| Molecular Weight (g/mol) | 166.1953 |
| Formula Weight | 166.0853 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction