Learn More
DL-Glutamic-2,4,4-d3 Acid, CDN
Product Code. 30321108
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30321108
Brand: CDN CDND11960.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material DL-Glutamic-d3 Acid
CAS 96927-56-9
Molecular Formula C5 D3 H6 N O4
Quantity 0.5 g
Synonym DL-Glutamic-2,4,4-d3 Acid, Glutamic-2,4,4-d3 acid (9 CI), DL-Glutamic-2,4,4-d3 acid (ZCI)
InChI Formula InChI=1 S/C5H9NO4/c6-3(5(9)10)1-2-4(7)8/h3H,1-2,6H2,(H,7,8)(H,9,10)/i2D2,3 D
SMILES [2 H]C([2 H])(CC([2 H])(N)C(=O)O)C(=O)O
IUPAC Name 2-amino-2,4,4-trideuteriopentanedioic acid
Molecular Weight (g/mol) 150.15
Formula Weight 150.072 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity