missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl Terephthalate-2,3,5,6-d4, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 1,4-Benzenedicarboxylic Acid-d4 Dimethyl Ester |
| CAS | 74097-01-9 |
| Molecular Formula | C10 D4 H6 O4 |
| Quantity | 0.5 g |
| Synonym | Terephthalic Acid-d4 Dimethyl Ester, DMT-d4, Dimethyl 1,4-Benzenedicarboxylate-d4, Dimethyl p-Benzenedicarboxylate-d4, Dimethyl p-Phthalate-d4, 1,4-Benzene-2,3,5,6-d4-dicarboxylic acid, dimethyl ester (9 CI), 1,4-Benzene-2,3,5,6-d4-dicarboxylic acid, dimethyl ester (9 CI), 1,4-Benzene-2,3,5,6-d4-dicarboxylic acid, dimethyl ester (9 CI), 1,4-Benzene-2,3,5,6-d4-dicarboxylic acid, dimethyl ester (9 CI), 1,4-Benzene-2,3,5,6-d4-dicarboxylic acid, dimethyl ester (9 CI), 1,4-Benzene-2,3,5,6-d4-dicarboxylic acid, dimethyl ester (9 CI), 1,4-Benzene-2,3,5,6-d4-dicarboxylic acid, dimethyl ester (9 CI), 1,4-Benzene-2,3,5,6-d4-dicarboxylic acid, dimethyl ester (9 CI), 1,4-Benzene-2,3,5,6-d4-dicarboxylic acid, dimethyl ester (9 CI), 1,4-Benzene-2,3,5,6-d4-dicarboxylic acid, dimethyl ester (9 CI), 1,4-Benzene-2,3,5,6-d4-dicarboxylic acid, dimethyl ester (9 CI) |
| InChI Formula | InChI=1 S/C10H10O4/c1-13-9(11)7-3-5-8(6-4-7)10(12)14-2/h3-6 H,1-2H3/i3D,4 D,5 D,6 D |
| SMILES | [2 H]c1c([2 H])c(C(=O)OC)c([2 H])c([2 H])c1C(=O)OC |
| IUPAC Name | dimethyl 2,3,5,6-tetradeuteriobenzene-1,4-dicarboxylate |
| Molecular Weight (g/mol) | 198.21 |
| Formula Weight | 198.083 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction