missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclopropyl-2,2,3,3-d4 Methyl Ketone, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 1-Cyclopropylethanone-2',2',3',3'-D4 |
| CAS | 350818-63-2 |
| Molecular Formula | C5 D4 H4 O |
| Quantity | 0.5 g |
| Synonym | Ethanone, 1-(cyclopropyl-2,2,3,3-d4)- (9 CI), 1-(Cyclopropyl-2,2,3,3-d4)ethanone, Cyclopropyl-2,2,3,3-d4 Methyl Ketone, 1-Cyclopropylethanone-D4, 1-Cyclopropylethanone-2',2',3',3'-D4 |
| InChI Formula | InChI=1 S/C5H8O/c1-4(6)5-2-3-5/h5H,2-3H2,1H3/i2D2,3D2 |
| SMILES | [2 H]C1([2 H])C(C(=O)C)C1([2 H])[2 H] |
| IUPAC Name | 1-(2,2,3,3-tetradeuteriocyclopropyl)ethanone |
| Molecular Weight (g/mol) | 88.1411 |
| Formula Weight | 88.0826 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction