missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromoethane-1,2,2-d3, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 1-Bromoethane-1,2,2-D3 |
| CAS | 23705-69-3 |
| Molecular Formula | C2 D3 H2 Br |
| Quantity | 0.5 g |
| Synonym | Ethane-1,1,2-d3, 2-bromo- (6 CI, 8 CI), 1-Bromoethane-1,2,2-D3, 2-Bromoethane-1,1,2-d3, Ethylbromide-1,2,2-d3, Bromoethane-d3 |
| InChI Formula | InChI=1 S/C2H5Br/c1-2-3/h2H2,1H3/i1D2,2 D |
| SMILES | [2 H]C([2 H])C([2 H])Br |
| IUPAC Name | 1-bromo-1,2,2-trideuterioethane |
| Molecular Weight (g/mol) | 111.9836 |
| Formula Weight | 110.9763 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction