Learn More
Acetanilide-d8, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Acetanilide-d8 |
| CAS | 22778-17-2 |
| Molecular Formula | C8 D8 H N O |
| Quantity | 0.5 g |
| Synonym | Acetamide-2,2,2-d3, N-(phenyl-2,3,4,5,6-d5)-, Acetamide-2,2,2-d3, N-(phenyl-d5)- (9 CI), Acetanilide-2,2,2,2',3',4',5',6'-d8 (8 CI), N-(Phenyl-2,3,4,5,6-d5)acetamide-2,2,2-d3, Acetanilide-d8, 2,2,2,2',3',4',5',6'-Octadeuteroacetanilide, 2,2,2,2',3',4',5',6'-Octadeuteroacetanilide, 2,2,2,2',3',4',5',6'-Octadeuteroacetanilide |
| InChI Formula | InChI=1 S/C8H9NO/c1-7(10)9-8-5-3-2-4-6-8/h2-6 H,1H3,(H,9,10)/i1D3,2 D,3 D,4 D,5 D,6 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(NC(=O)C([2 H])([2 H])[2 H])c([2 H])c1[2 H] |
| IUPAC Name | 2,2,2-trideuterio-N-(2,3,4,5,6-pentadeuteriophenyl)acetamide |
| Molecular Weight (g/mol) | 143.213 |
| Formula Weight | 143.119 g/mol |
| Show More |