missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetaldehyde-2,2,2-d3, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Acetaldehyde-d3 |
| CAS | 19901-15-6 |
| Molecular Formula | C2 D3 H O |
| Quantity | 0.5 g |
| Synonym | Acetic Aldehyde-d3, Ethanal-d3, Ethyl Aldehyde-d3, Perdeuteroacetaldehyde-d3, Tetradeuteroacetaldehyde-d3, Acetaldehyde-2,2,2-d3 |
| InChI Formula | InChI=1 S/C2H4O/c1-2-3/h2H,1H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])C=O |
| IUPAC Name | 2,2,2-trideuterioacetaldehyde |
| Molecular Weight (g/mol) | 47.07 |
| Formula Weight | 47.045 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction