Learn More
4'-Methyl-d3-acetophenone, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 4'-Methylacetophenone 4'-methyl-d3 |
| CAS | 114379-92-9 |
| Molecular Formula | C9 2H3 H7 O |
| Quantity | 0.5 g |
| Synonym | Ethanone, 1-[4-(methyl-d3)phenyl]-, Acetophenone, 4'-methyl-d3- (6 CI), 1-[4-(Methyl-d3)phenyl]ethanone, (4-(Trideuteriomethyl)phenyl)ethan-1-one, 1-(4-Trideuteriomethyl-phenyl)-ethanone, 4'-Methylacetophenone-d3, 4'-Methylacetophenone-d3, 4'-Methylacetophenone-d3 |
| InChI Formula | InChI=1 S/C9H10O/c1-7-3-5-9(6-4-7)8(2)10/h3-6 H,1-2H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])c1ccc(cc1)C(=O)C |
| IUPAC Name | 1-[4-(trideuteriomethyl)phenyl]ethanone |
| Molecular Weight (g/mol) | 137.19 |
| Formula Weight | 137.092 g/mol |
| Show More |