Learn More
4-Bromochlorobenzene-d4, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 4-Bromochlorobenzene-D4 |
| CAS | 134415-42-2 |
| Molecular Formula | C6 D4 Br Cl |
| Quantity | 0.5 g |
| Synonym | Benzene-1,2,4,5-d4, 3-bromo-6-chloro- (9 CI), 4-Bromochlorobenzene-d4, 4-Bromochlorobenzene-2,3,5,6-d4, 4-Bromochlorobenzene-D4, 3-Bromo-6-chlorobenzene-1,2,4,5-d4 |
| InChI Formula | InChI=1 S/C6H4BrCl/c7-5-1-3-6(8)4-2-5/h1-4 H/i1D,2 D,3 D,4 D |
| SMILES | [2 H]c1c([2 H])c(Br)c([2 H])c([2 H])c1Cl |
| IUPAC Name | 1-bromo-4-chloro-2,3,5,6-tetradeuteriobenzene |
| Molecular Weight (g/mol) | 195.4776 |
| Formula Weight | 193.9436 g/mol |
| Show More |