Learn More
4,4'-Dipyridyl-d8, CDN
Product Code. 30319623
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30319623
Brand: CDN CDND50730.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4,4'-Bipyridine-d8
CAS 132125-39-4
Molecular Formula C10 2H8 N2
Quantity 0.5 g
Synonym 4,4'-Bipyridine-2,2',3,3',5,5',6,6'-d8, 4,4'-Bipyridine-d8, SERS 421
InChI Formula InChI=1 S/C10H8N2/c1-5-11-6-2-9(1)10-3-7-12-8-4-10/h1-8 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D
SMILES [2 H]c1nc([2 H])c([2 H])c(c1[2 H])c2c([2 H])c([2 H])nc([2 H])c2[2 H]
IUPAC Name 2,3,5,6-tetradeuterio-4-(2,3,5,6-tetradeuteriopyridin-4-yl)pyridine
Molecular Weight (g/mol) 164.23
Formula Weight 164.119 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity