Learn More
3-Fluorotoluene-alpha-d1, CDN
Product Code. 30325216
Click to view available options
Quantity:
0.25 g
0.5 g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30325216
Brand: CDN CDND76260.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-Fluorotoluene-alpha-D
CAS 2118336-62-0
Molecular Formula C7 D H6 F
Quantity 0.5 g
Synonym 1-Fluoro-3-(methyl-d)benzene, 3-Fluorotoluene-α-d1, 3-Fluorotoluene-α-D
InChI Formula InChI=1 S/C7H7F/c1-6-3-2-4-7(8)5-6/h2-5 H,1H3/i1D
SMILES [2 H]Cc1cccc(F)c1
IUPAC Name 1-(deuteriomethyl)-3-fluorobenzene
Molecular Weight (g/mol) 111.135
Formula Weight 111.0595 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity