Learn More
2-Nitrotoluene-d7, CDN
Product Code. 30310095
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30310095
Brand: CDN CDND76890.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Nitrotoluene-d7
CAS 84344-04-7
Molecular Formula C7 2H7 N O2
Quantity 0.5 g
Synonym 2-Nitrotoluene-d7
InChI Formula InChI=1 S/C7H7NO2/c1-6-4-2-3-5-7(6)8(9)10/h2-5 H,1H3/i1D3,2 D,3 D,4 D,5 D
SMILES [2 H]c1c([2 H])c([2 H])c(c(c1[2 H])[N+](=O)[O-])C([2 H])([2 H])[2 H]
IUPAC Name 1,2,3,4-tetradeuterio-5-nitro-6-(trideuteriomethyl)benzene
Molecular Weight (g/mol) 144.18
Formula Weight 144.0916 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity