missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methylpyridine-d7, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 2-Picoline-d7 |
| CAS | 93951-93-0 |
| Molecular Formula | C6 D7 N |
| Quantity | 0.5 g |
| Synonym | 2-Methylpyridine-d7, o-Picoline-d7, o-Methylpyridine-d7, ortho-Picoline-d7, ortho-Methylpyridine-d7 |
| InChI Formula | InChI=1 S/C6H7N/c1-6-4-2-3-5-7-6/h2-5 H,1H3/i1D3,2 D,3 D,4 D,5 D |
| SMILES | [2 H]c1nc(c([2 H])c([2 H])c1[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 2,3,4,5-tetradeuterio-6-(trideuteriomethyl)pyridine |
| Molecular Weight (g/mol) | 100.17 |
| Formula Weight | 100.1018 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction