Learn More
2-Methyl-d3-pyridine, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 2-Methyl-d3-pyridine |
| CAS | 10259-19-5 |
| Molecular Formula | C6 D3 H4 N |
| Quantity | 0.5 g |
| Synonym | Pyridine, 2-(methyl-d3)- (9 CI), 2-Picoline-α,α,α-d3 (7 CI, 8 CI), 2-(Methyl-d3)pyridine, 2-Methylpyridine-d3, 2-Methylpyridine-D3 (methyl-D3), 2-Picoline-d3, 2-Methylpyridine-α,α,α-D3 |
| InChI Formula | InChI=1 S/C6H7N/c1-6-4-2-3-5-7-6/h2-5 H,1H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])c1ccccn1 |
| IUPAC Name | 2-(trideuteriomethyl)pyridine |
| Molecular Weight (g/mol) | 96.145 |
| Formula Weight | 96.0767 g/mol |
| Show More |