Learn More
2-Ethyl-6-methylaniline-d13, CDN
Product Code. 30320818
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30320818
Brand: CDN CDND59190.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Ethyl-6-methylaniline-d13
CAS 1219794-93-0
Molecular Formula C9 D13 N
Quantity 0.5 g
Synonym 2-(Ethyl-d5)-6-(methyl-d3)benzen-3,4,5-d3-amine-d2, 2-Ethyl-6-methylaniline-d13, 2-Ethyl-6-methylaniline-D13
InChI Formula InChI=1 S/C9H13N/c1-3-8-6-4-5-7(2)9(8)10/h4-6 H,3,10H2,1-2H3/i1D3,2D3,3D2,4 D,5 D,6 D/hD2
SMILES [2 H]N([2 H])c1c(c([2 H])c([2 H])c([2 H])c1C([2 H])([2 H])C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H]
IUPAC Name N,N,3,4,5-pentadeuterio-2-(1,1,2,2,2-pentadeuterioethyl)-6-(trideuteriomethyl)aniline
Molecular Weight (g/mol) 148.2863
Formula Weight 148.1864 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity