Learn More
2-Chloropropane-1,1,1-d3, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 2-Chloropropane-1,1,1-D3 |
| CAS | 137832-55-4 |
| Molecular Formula | C3 D3 H4 Cl |
| Quantity | 0.5 g |
| Synonym | Propane-1,1,1-d3, 2-chloro- (9 CI), 2-Chloropropane-1,1,1-d3, 2-Chloro-1,1,1-trideuteriopropane, Isopropyl-d3 chloride, 2-Chloropropane-d3, 2-Chloropropane-1,1,1-D3 |
| InChI Formula | InChI=1 S/C3H7Cl/c1-3(2)4/h3H,1-2H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])C(C)Cl |
| IUPAC Name | 2-chloro-1,1,1-trideuteriopropane |
| Molecular Weight (g/mol) | 81.5592 |
| Formula Weight | 81.0425 g/mol |
| Show More |