Learn More
2-Bromopyridine-d4, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 2-Bromopyridine-d4 |
| CAS | 70766-71-1 |
| Molecular Formula | C5 2H4 Br N |
| Quantity | 0.5 g |
| InChI Formula | InChI=1 S/C5H4BrN/c6-5-3-1-2-4-7-5/h1-4 H/i1D,2 D,3 D,4 D |
| SMILES | [2 H]c1nc(Br)c([2 H])c([2 H])c1[2 H] |
| IUPAC Name | 2-bromo-3,4,5,6-tetradeuteriopyridine |
| Molecular Weight (g/mol) | 162.02 |
| Formula Weight | 160.9778 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |