missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dichlorophenol-OD, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.5g
Specifications
Specifications
| Chemical Name or Material | 2,3-Dichlorophenol-OD |
| CAS | 59457-02-2 |
| Molecular Formula | C6 D H3 Cl2 O |
| Quantity | 0.5 g |
| Synonym | Phenol-d, 2,3-dichloro- (9 CI), 2,3-Dichlorophenol-OD, 2,3-Dichlorophenol-d |
| InChI Formula | InChI=1 S/C6H4Cl2O/c7-4-2-1-3-5(9)6(4)8/h1-3,9 H/i/hD |
| SMILES | [2 H]Oc1cccc(Cl)c1Cl |
| IUPAC Name | 1,2-dichloro-3-deuteriooxybenzene |
| Molecular Weight (g/mol) | 164.0075 |
| Formula Weight | 162.9702 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction