Learn More
(±)-1-Phenylethan-2,2,2-d3-ol, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 1-Phenylethanol-2,2,2-D3 |
| CAS | 17537-32-5 |
| Molecular Formula | C8 D3 H7 O |
| Quantity | 0.5 g |
| Synonym | Benzenemethanol, α-(methyl-d3)- (9 CI, ACI), α-(Methyl-d3)benzenemethanol (ACI), 2,2,2-Trideuterio-1-phenylethanol, 1-Phenylethanol-2,2,2-d3, 1-Phenylethanol-2,2,2-D3, (±)-1-Phenylethan-2,2,2-d3-ol, (±)-1-Phenylethan-2,2,2-d3-ol |
| InChI Formula | InChI=1 S/C8H10O/c1-7(9)8-5-3-2-4-6-8/h2-7,9 H,1H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])C(O)c1ccccc1 |
| IUPAC Name | 2,2,2-trideuterio-1-phenylethanol |
| Molecular Weight (g/mol) | 125.1829 |
| Formula Weight | 125.092 g/mol |
| Show More |