Learn More
1-Iodopropane-1,1-d2, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 1-Iodopropane-1,1-D2 |
| CAS | 25493-14-5 |
| Molecular Formula | C3 D2 H5 I |
| Quantity | 0.5 g |
| Synonym | Propane-1,1-d2, 1-iodo- (6 CI, 8 CI, 9 CI), 1-Iodopropane-d2, 1-Iodopropane-1,1-d2, 1-Iodopropane-1,1-D2, [1,1-2H2]Propyl iodide |
| InChI Formula | InChI=1 S/C3H7I/c1-2-3-4/h2-3H2,1H3/i3D2 |
| SMILES | [2 H]C([2 H])(I)CC |
| IUPAC Name | 1,1-dideuterio-1-iodopropane |
| Molecular Weight (g/mol) | 172.0045 |
| Formula Weight | 171.9718 g/mol |
| Show More |