missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromododecane-d25, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 1-Bromododecane-D25 |
| CAS | 204259-66-5 |
| Molecular Formula | C12 D25 Br |
| Quantity | 0.5 g |
| Synonym | Dodecane-1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12-d25, 12-bromo- (9 CI, ACI), 12-Bromododecane-1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12-d25 (ACI), 1-Bromododecane-d25, 1-Bromododecane-D25, Dodecyl bromide-d25, Lauryl bromide-d25 |
| InChI Formula | InChI=1 S/C12H25Br/c1-2-3-4-5-6-7-8-9-10-11-12-13/h2-12H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])Br |
| IUPAC Name | 1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-pentacosadeuteriododecane |
| Molecular Weight (g/mol) | 274.3849 |
| Formula Weight | 273.2709 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction