missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Dimethoxybenzene-4,5-d2, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 1,2-Dimethoxybenzene-4,5-D2 |
| CAS | 203645-56-1 |
| Molecular Formula | C8 D2 H8 O2 |
| Quantity | 0.5 g |
| Synonym | Benzene-1,2-d2, 4,5-dimethoxy- (9 CI), 4,5-Dimethoxybenzene-1,2-d4, 1,2-Dimethoxybenzene-4,5-D2, 1,2-Dimethoxybenzene-d2, 2-Methoxyanisole-4,5-d2, Catechol-d2 dimethyl ether, o-Hydroquinone-d2 dimethyl ether |
| InChI Formula | InChI=1 S/C8H10O2/c1-9-7-5-3-4-6-8(7)10-2/h3-6 H,1-2H3/i3D,4 D |
| SMILES | [2 H]c1cc(OC)c(OC)cc1[2 H] |
| IUPAC Name | 1,2-dideuterio-4,5-dimethoxybenzene |
| Molecular Weight (g/mol) | 140.176 |
| Formula Weight | 140.081 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction