missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Benzenediamine-d8, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 1,2-Phenylenediamine-d8 |
| CAS | 1219798-78-3 |
| Molecular Formula | C6D8N2 |
| Quantity | 0.5 g |
| Synonym | 1,2-Benzenediamine-d8, o-Phenylenediamine-d8, 1,2-Diaminobenzene-d8, 2-Aminoaniline-d8, C.I. 76010-d8, C.I. Oxidation Base 16-d8, IK 3, NSC 5354-d8, Orthamine-d8, o-Aminoaniline-d8, o-Aminophenylamine-d8, o-Benzenediamine-d8, o-Diaminobenzene-d8 |
| InChI Formula | InChI=1 S/C6H8N2/c7-5-3-1-2-4-6(5)8/h1-4 H,7-8H2/i1D,2 D,3 D,4 D/hD4 |
| SMILES | [2 H]C1=C(N([2 H])[2 H])C(N([2 H])[2 H])=C([2 H])C([2 H])=C1[2 H] |
| IUPAC Name | benzene-3,4,5,6-d4-1,2-diamine-N1,N1,N2,N2-d4 |
| Molecular Weight (g/mol) | 116.19 |
| Formula Weight | 116.12 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction