Learn More
1,2,3,4-Tetramethylbenzene-d14, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 1,2,3,4-Tetramethylbenzene-D14 |
| CAS | 350818-60-9 |
| Molecular Formula | C10 D14 |
| Quantity | 0.5 g |
| Synonym | Benzene-1,2-d2, 3,4,5,6-tetra(methyl-d3)- (9 CI), 3,4,5,6-Tetra(methyl-d3)benzene-1,2-d2, 1,2,3,4-Tetramethylbenzene-D14, 1,2,3,4-Tetramethylbenzene-d14, Prehnitene-d14, Prehnitol-d14 |
| InChI Formula | InChI=1 S/C10H14/c1-7-5-6-8(2)10(4)9(7)3/h5-6 H,1-4H3/i1D3,2D3,3D3,4D3,5 D,6 D |
| SMILES | [2 H]c1c([2 H])c(c(c(c1C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,2-dideuterio-3,4,5,6-tetrakis(trideuteriomethyl)benzene |
| Molecular Weight (g/mol) | 148.3044 |
| Formula Weight | 148.1974 g/mol |
| Show More |