missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethanolamine-d15, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Triethanolamine-D15 |
| CAS | 73205-32-0 |
| Molecular Formula | C6 2H15 N O3 |
| Quantity | 0.25 g |
| InChI Formula | InChI=1 S/C6H15NO3/c8-4-1-7(2-5-9)3-6-10/h8-10 H,1-6H2/i1D2,2D2,3D2,4D2,5D2,6D2,8 D,9 D,10 D |
| SMILES | [2 H]OC([2 H])([2 H])C([2 H])([2 H])N(C([2 H])([2 H])C([2 H])([2 H])O[2 H])C([2 H])([2 H])C([2 H])([2 H])O[2 H] |
| IUPAC Name | 1,1,2,2-tetradeuterio-2-deuteriooxy-N,N-bis(1,1,2,2-tetradeuterio-2-deuteriooxyethyl)ethanamine |
| Molecular Weight (g/mol) | 164.28 |
| Formula Weight | 164.1993 g/mol |
| Percent Purity | 98 atom % D, min 97% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction