missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetradecanoic-12,12,13,13,14,14,14-d7 Acid, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Tetradecanoic-12,12,13,13,14,14,14-D7 Acid |
| Molecular Formula | C14 D7 H21 O2 |
| Quantity | 0.25 g |
| Synonym | Tetradecanoic-12,12,13,13,14,14,14-d7 acid, Myristic acid-12,12,13,13,14,14,14-d7 |
| InChI Formula | InChI=1 S/C14H28O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14(15)16/h2-13H2,1H3,(H,15,16)/i1D3,2D2,3D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])CCCCCCCCCCC(=O)O |
| IUPAC Name | 12,12,13,13,14,14,14-heptadeuteriotetradecanoic acid |
| Molecular Weight (g/mol) | 235.414 |
| Formula Weight | 235.253 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction